1-(2,2-Diethoxyethyl)-1H-pyrazole-4-boronic acid ≥98%

Supplier: Ambeed
Warning

A190189-100MG A190189-1G A190189-250MG
77901-170EA 47 USD
77901-170 77901-109 77901-627
1-(2,2-Diethoxyethyl)-1H-pyrazole-4-boronic acid ≥98%
1-(2,2-Diethoxyethyl)-1H-pyrazole-4-boronic acid

Precaución: for Research Use Only

Formula: C₉H₁₇BN₂O₄
MDL Number: MFCD12756418
CAS Number: 1217501-20-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR