4-tert-Octylphenol ≥95%

Supplier: Ambeed
Danger

Synonyms: 4-tert-Octylphenol, 4-(1,1,3,3-Tetramethylbutyl)phenol

A190172-25G A190172-1000G A190172-100G
77287-468EA 7.21 USD
77287-468 77615-608 77615-610
4-tert-Octylphenol ≥95%
4-tert-Octylphenol

Precaución: for Research Use Only

Formula: C₁₄H₂₂O
MW: 206.33 g/mol
MDL Number: MFCD00002368
CAS Number: 140-66-9
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR