4-Butylphenylboronic acid pinacol ester ≥95%

Supplier: Ambeed
Warning

Synonyms: 2-(4-Butylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

A189356-1G A189356-5G
77685-448EA 79 USD
77685-448 77685-449
4-Butylphenylboronic acid pinacol ester ≥95%
4-Butylphenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₆H₂₅BO₂
MDL Number: MFCD20527105
CAS Number: 625458-85-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR