Boc-β-chloro-Ala-OH ≥95%

Supplier: Ambeed
Warning

A189236-5G A189236-1G
77671-680EA 70 USD
77671-680 77881-410
Boc-β-chloro-Ala-OH ≥95%
Boc-beta-chloro-Ala-OH

Precaución: for Research Use Only

Formula: C₈H₁₄ClNO₄
MDL Number: MFCD00672318
CAS Number: 71404-98-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR