(R)-(+)-Methyl p-tolyl sulfoxide ≥98%

Supplier: Ambeed
Warning

Synonyms: (R)-1-Methyl-4-(methylsulphinyl)benzene

A189124-1G A189124-100MG
77746-214EA 130 USD
77746-214 77746-213
(R)-(+)-Methyl p-tolyl sulfoxide ≥98%
(R)-(+)-Methyl p-tolyl sulfoxide

Precaución: for Research Use Only

Formula: C₈H₁₀OS
MW: 154.229 g/mol
MDL Number: MFCD00151502
CAS Number: 1519-39-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR