Boc-(S)-3-amino-4-(4-nitrophenyl)butyric acid ≥98%

Supplier: Ambeed
Warning

A188944-1G A188944-250MG
77871-947EA 79 USD
77871-947 77871-948
Boc-(S)-3-amino-4-(4-nitrophenyl)butyric acid ≥98%
Boc-(S)-3-amino-4-(4-nitrophenyl)butyric acid

Precaución: for Research Use Only

Formula: C₁₅H₂₀N₂O₆
MDL Number: MFCD01076286
CAS Number: 127106-71-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR