6-Chloropyridazine-3,4-diamine ≥95%

Supplier: Ambeed
Warning

A188621-250MG A188621-1G
77611-826EA 155.53 USD
77611-826 77890-166
6-Chloropyridazine-3,4-diamine ≥95%
6-Chloropyridazine-3,4-diamine

Precaución: for Research Use Only

Formula: C₄H₅ClN₄
CAS Number: 932-50-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR