Ac-rC Phosphoramidite ≥95%

Supplier: Ambeed
Warning

A187780-100MG A187780-250MG A187780-5G A187780-1G A187780-25G
77671-662EA 31 USD
77671-662 77671-664 77671-666 77881-404 77881-405
Ac-rC Phosphoramidite ≥95%
Ac-rC Phosphoramidite

Precaución: for Research Use Only

Formula: C₄₇H₆₄N₅O₉PSi
MW: 902.09 g/mol
MDL Number: MFCD09842111
CAS Number: 121058-88-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR