1-(4-Boronophenyl)cyclopropanecarboxylic acid ≥98%

Supplier: Ambeed
Warning

A187341-250MG A187341-1G A187341-5G
77671-656EA 89 USD
77671-656 77883-397 77883-398
1-(4-Boronophenyl)cyclopropanecarboxylic acid ≥98%
1-(4-Boronophenyl)cyclopropanecarboxylic acid
Formula: C₁₀H₁₁BO₄
MDL Number: MFCD09037835
CAS Number: 1159489-46-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR