3-Chlorothiophene-2-boronic acid pinacol ester 95+%

Supplier: Ambeed
Warning

Synonyms: 2-(3-Chlorothiophen-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

A187218-100MG A187218-1G A187218-250MG
77671-652EA 80 USD
77671-652 77883-394 77883-395
3-Chlorothiophene-2-boronic acid pinacol ester 95+%
3-Chlorothiophene-2-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₀H₁₄BClO₂S
MDL Number: MFCD09837624
CAS Number: 1040281-97-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR