Boc-D-Dab(Fmoc)-OH ≥98%

Supplier: Ambeed
Warning

A186848-1G A186848-250MG
77600-518EA 18.45 USD
77600-518 77871-929
Boc-D-Dab(Fmoc)-OH ≥98%
Boc-D-Dab(Fmoc)-OH

Precaución: for Research Use Only

Formula: C₂₄H₂₈N₂O₆
MW: 440.496 g/mol
MDL Number: MFCD00798625
CAS Number: 131570-57-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR