5,5'-Dichloro-[1,1'-biphenyl]-2,2'-diamine 95+%

Supplier: Ambeed
Warning

A1867642-100MG A1867642-1G A1867642-250MG
77877-375EA 46 USD
77877-375 77877-376 77877-377
5,5'-Dichloro-[1,1'-biphenyl]-2,2'-diamine 95+%
5,5'-Dichloro-[1,1'-biphenyl]-2,2'-diamine

Precaución: for Research Use Only

Formula: C₁₂H₁₀Cl₂N₂
CAS Number: 2849-36-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR