4-Iodo-m-toluidine ≥97%

Supplier: Ambeed
Warning

A186309-100G A186309-10G
77885-411EA 360 USD
77885-411 77885-412
4-Iodo-m-toluidine ≥97%
4-Iodo-m-toluidine

Precaución: for Research Use Only

Formula: C₇H₈IN
MDL Number: MFCD01569451
CAS Number: 4949-69-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR