(1S,2S)-2-(2-((3-(1H-Benzo[d]imidazol-2-yl)propyl)(methyl)amino)ethyl)-6-fluoro-1-isopropyl-1,2,3,4-tetrahydronaphthalen-2-yl 2-methoxyacetate dihydrochloride ≥97%

Supplier: Ambeed
Warning

Synonyms: Mibefradil 2HCl

A186181-10MG A186181-1MG A186181-5MG
77600-504EA 868.18 USD
77600-504 77600-506 77600-508
(1S,2S)-2-(2-((3-(1H-Benzo[d]imidazol-2-yl)propyl)(methyl)amino)ethyl)-6-fluoro-1-isopropyl-1,2,3,4-tetrahydronaphthalen-2-yl 2-methoxyacetate dihydrochloride ≥97%
(1S,2S)-2-(2-((3-(1H-Benzo[d]imidazol-2-yl)propyl)(methyl)amino)ethyl)-6-fluoro-1-isopropyl-1,2,3,4-tetrahydronaphthalen-2-yl 2-methoxyacetate dihydrochloride

Precaución: for Research Use Only

Formula: C₂₉H₄₀Cl₂FN₃O₃
MDL Number: MFCD00881804
CAS Number: 116666-63-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR