3-Bromo-4,5-dihydroxybenzaldehyde ≥95%

Supplier: Ambeed
Warning

A185736-1G A185736-250MG
77625-884EA 103 USD
77625-884 77625-886
3-Bromo-4,5-dihydroxybenzaldehyde ≥95%
3-Bromo-4,5-dihydroxybenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₅BrO₃
MDL Number: MFCD00016608
CAS Number: 16414-34-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR