rel-tert-Butyl ((1S,2S)-2-hydroxycyclobutyl)carbamate ≥97%

Supplier: Ambeed
Warning

A184468-1G A184468-250MG A184468-5G
77615-560EA 938.33 USD
77615-560 77615-562 77615-564
rel-tert-Butyl ((1S,2S)-2-hydroxycyclobutyl)carbamate ≥97%
rel-tert-Butyl ((1S,2S)-2-hydroxycyclobutyl)carbamate

Precaución: for Research Use Only

Formula: C₉H₁₇NO₃
MW: 187.24 g/mol
MDL Number: MFCD09952307
CAS Number: 1354422-81-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR