5-Fluoroquinoline-8-boronic acid ≥98%

Supplier: Ambeed
Warning

Synonyms: (5-Fluoroquinolin-8-yl)boronic acid

A184196-1G A184196-250MG
77885-405EA 282 USD
77885-405 77885-406
5-Fluoroquinoline-8-boronic acid ≥98%
5-Fluoroquinoline-8-boronic acid

Precaución: for Research Use Only

Formula: C₉H₇BFNO₂
MDL Number: MFCD11504850
CAS Number: 1072951-45-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR