N,N',N''-Triacetylchitotriose ≥98%

Supplier: Ambeed
Warning

Synonyms: Tri-N-acetylchitotriose, N-((2S,3R,4R,5S,6R)-2-(((2R,3S,4R,5R)-5-Acetamido-1,2,4-trihydroxy-6-oxohexan-3-yl)oxy)-5-(((2S,3R,4R,5S,6R)-3-acetamido-4,5-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-4-hydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-3-yl)acetamide, GlcNAc beta(1-4)GlcNAc beta(1-4)GlcNAc

A184187-10MG A184187-5MG A184187-50MG
77600-484EA 177.33 USD
77600-484 77600-486 77879-191
N,N',N''-Triacetylchitotriose ≥98%
N,N',N''-Triacetylchitotriose

Precaución: for Research Use Only

Formula: C₂₄H₄₁N₃O₁₆
MDL Number: MFCD00136047
CAS Number: 38864-21-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR