3-Cyano-5-methylphenylboronic acid, pinacol ester ≥95%

Supplier: Ambeed
Warning

Synonyms: 3-Cyano-5-methylphenylboronic acid pinacol ester, 3-Methyl-5-(4,4,5,5-tetramethyl[1,3,2]dioxaborolan-2-yl)benzonitrile

A183857-250MG A183857-5G A183857-100MG A183857-1G
77593-182EA 36.9 USD
77593-182 77611-786 77671-596 77671-598
3-Cyano-5-methylphenylboronic acid, pinacol ester ≥95%
3-Cyano-5-methylphenylboronic acid, pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₁₈BNO₂
MW: 243.109 g/mol
MDL Number: MFCD18383798
CAS Number: 1220219-59-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR