3-((4-((5-Hydroxy-2-methylphenyl)amino)pyrimidin-2-yl)amino)benzamide ≥98%

Supplier: Ambeed
Warning

Synonyms: Lck inhibitor 2

A183410-10MG A183410-5MG
77746-189EA 99 USD
77746-189 77746-190
3-((4-((5-Hydroxy-2-methylphenyl)amino)pyrimidin-2-yl)amino)benzamide ≥98%
3-((4-((5-Hydroxy-2-methylphenyl)amino)pyrimidin-2-yl)amino)benzamide

Precaución: for Research Use Only

Formula: C₁₈H₁₇N₅O₂
MW: 335.36 g/mol
MDL Number: MFCD18206813
CAS Number: 944795-06-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR