4-(2-Phenyl-1H-benzimidazol-1-yl)phenylboronic acid ≥95%

Supplier: Ambeed
Warning

Synonyms: (4-(2-Phenyl-1H-benzo[d]imidazol-1-yl)phenyl)boronic acid, 4-(2-Phenyl-1H-benzimidazol-1-yl)benzeneboronic acid

A182207-1G A182207-100MG A182207-250MG A182207-5G
77671-576EA 133 USD
77671-576 77883-375 77883-376 77883-377
4-(2-Phenyl-1H-benzimidazol-1-yl)phenylboronic acid ≥95%
4-(2-Phenyl-1H-benzimidazol-1-yl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₉H₁₅BN₂O₂
MDL Number: MFCD11977298
CAS Number: 867044-33-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR