2,5-Dioxopyrrolidin-1-yl 1-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)-3-oxo-7,10,13-trioxa-4-azahexadecan-16-oate ≥98%

Supplier: Ambeed
Warning

Synonyms: Mal-amido-PEG3-NHS ester

A181555-100MG A181555-250MG
77748-187EA 53 USD
77748-187 77748-188
2,5-Dioxopyrrolidin-1-yl 1-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)-3-oxo-7,10,13-trioxa-4-azahexadecan-16-oate ≥98%
2,5-Dioxopyrrolidin-1-yl 1-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)-3-oxo-7,10,13-trioxa-4-azahexadecan-16-oate

Precaución: for Research Use Only

Formula: C₂₀H₂₇N₃O₁₀
MW: 469.44 g/mol
CAS Number: 2055353-77-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR