4-Methoxy-5-(3-morpholinopropoxy)-2-nitrobenzonitrile ≥97%

Supplier: Ambeed
Warning

Synonyms: Gefitinib impurity 1

A181163-1G A181163-5G
77611-756EA 16.48 USD
77611-756 77611-758
4-Methoxy-5-(3-morpholinopropoxy)-2-nitrobenzonitrile ≥97%
4-Methoxy-5-(3-morpholinopropoxy)-2-nitrobenzonitrile

Precaución: for Research Use Only

Formula: C₁₅H₁₉N₃O₅
MDL Number: MFCD11110476
CAS Number: 675126-26-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR