5-Fluoro-2-(trifluoromethoxy)aniline ≥98%

Supplier: Ambeed
Warning

Synonyms: 5-Fluoro-2-(trifluoromethoxy)benzylamine

A180909-5G A180909-100G A180909-1G A180909-25G
77685-417EA 52 USD
77685-417 77900-540 77902-003 77901-555
5-Fluoro-2-(trifluoromethoxy)aniline ≥98%
5-Fluoro-2-(trifluoromethoxy)aniline

Precaución: for Research Use Only

Formula: C₇H₅F₄NO
MDL Number: MFCD16652514
CAS Number: 123572-63-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR