(2,5-Bis(methoxycarbonyl)phenyl)boronic acid ≥98%

Supplier: Ambeed
Warning

A1809024-100MG A1809024-1G A1809024-250MG
77877-310EA 47 USD
77877-310 77877-311 77877-312
(2,5-Bis(methoxycarbonyl)phenyl)boronic acid ≥98%
(2,5-Bis(methoxycarbonyl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁BO₆
MW: 238 g/mol
MDL Number: MFCD23379492
CAS Number: 1374689-20-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR