(5-Carbamoyl-2-methylphenyl)boronic acid ≥98%

Supplier: Ambeed
Warning

A1807966-1G A1807966-250MG
77877-291EA 210 USD
77877-291 77877-292
(5-Carbamoyl-2-methylphenyl)boronic acid ≥98%
(5-Carbamoyl-2-methylphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₈H₁₀BNO₃
MDL Number: MFCD28147727
CAS Number: 1156500-84-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR