2-(3-Bromo-5-(tert-butyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥95%

Supplier: Ambeed
Warning

A1806177-1G A1806177-250MG A1806177-5G
77877-282EA 210 USD
77877-282 77877-283 77877-284
2-(3-Bromo-5-(tert-butyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥95%
2-(3-Bromo-5-(tert-butyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₆H₂₄BBrO₂
MW: 339.08 g/mol
MDL Number: MFCD29094852
CAS Number: 1644527-00-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR