(2S,4R)-1-((S)-2-(6-Aminohexanamido)-3,3-dimethylbutanoyl)-4-hydroxy-N-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide hydrochloride ≥95%
Supplier: Ambeed
|
Warning
|
A1805832-100MG
A1805832-250MG
77598-240EA
288.03
USD
77598-240
77625-854
(2S,4R)-1-((S)-2-(6-Aminohexanamido)-3,3-dimethylbutanoyl)-4-hydroxy-N-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide hydrochloride ≥95%
(2S,4R)-1-((S)-2-(6-Aminohexanamido)-3,3-dimethylbutanoyl)-4-hydroxy-N-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide hydrochloride
Precaución: for Research Use Only
Formula:
C₂₈H₄₂ClN₅O₄S MW: 580.18 g/mol |
CAS Number:
2170695-20-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...