1a,9b-Dihydrooxireno[2,3-f][1,10]phenanthroline ≥95%

Supplier: Ambeed
Warning

A180425-1G A180425-250MG A180425-5G
77877-277EA 115 USD
77877-277 77877-278 77877-279
1a,9b-Dihydrooxireno[2,3-f][1,10]phenanthroline ≥95%
1a,9b-Dihydrooxireno[2,3-f][1,10]phenanthroline

Precaución: for Research Use Only

Formula: C₁₂H₈N₂O
MDL Number: MFCD04039972
CAS Number: 65115-91-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR