3-Fluoro-4-(2-fluorophenoxy)aniline ≥97%

Supplier: Ambeed
Warning

A180372-1G A180372-100MG A180372-250MG A180372-5G
77600-456EA 27.68 USD
77600-456 77881-367 77881-368 77881-369
3-Fluoro-4-(2-fluorophenoxy)aniline ≥97%
3-Fluoro-4-(2-fluorophenoxy)aniline

Precaución: for Research Use Only

Formula: C₁₂H₉F₂NO
MDL Number: MFCD08687814
CAS Number: 946699-01-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR