8-Chlorochroman-4-one ≥95%

Supplier: Ambeed
Warning

A179824-1G A179824-5G
77625-852EA 17.51 USD
77625-852 77877-270
8-Chlorochroman-4-one ≥95%
8-Chlorochroman-4-one

Precaución: for Research Use Only

Formula: C₉H₇ClO₂
MDL Number: MFCD03695451
CAS Number: 49701-11-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR