4-(Difluoromethoxy)-3-hydroxybenzoic acid ≥95%

Supplier: Ambeed
Warning

A179765-100MG A179765-1G A179765-250MG
77890-145EA 15 USD
77890-145 77890-146 77890-147
4-(Difluoromethoxy)-3-hydroxybenzoic acid ≥95%
4-(Difluoromethoxy)-3-hydroxybenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆F₂O₄
MDL Number: MFCD24497448
CAS Number: 913985-07-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR