6-Bromo-N-methylpyridazin-3-amine ≥95%, Ambeed.Inc
Supplier: Ambeed
|
Warning
|
A179233-1G
A179233-250MG
77683-436EA
49
USD
77683-436
77683-437
6-Bromo-N-methylpyridazin-3-amine ≥95%, Ambeed.Inc
6-Bromo-N-methylpyridazin-3-amine
Precaución: For research use only.
Formula:
C₅H₆BrN₃ MW: 188.03 g/mol |
MDL Number:
MFCD13188585 CAS Number: 1208657-07-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...