5-Chloro-3-(trifluoromethyl)pyridin-2-amine ≥95%, Ambeed.Inc
Supplier: Ambeed
|
Warning
|
A179190-5G
A179190-1G
77683-435EA
43
USD
77683-435
77885-380
5-Chloro-3-(trifluoromethyl)pyridin-2-amine ≥95%, Ambeed.Inc
5-Chloro-3-(trifluoromethyl)pyridin-2-amine
Precaución: For research use only.
Formula:
C₆H₄ClF₃N₂ |
MDL Number:
MFCD13185329 CAS Number: 79456-33-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...