Citicoline ≥96%

Supplier: Ambeed
Warning

Synonyms: Cytidine 5'-diphosphocholine, 2-(((((((2R,3S,4R,5R)-5-(4-Amino-2-oxopyrimidin-1(2H)-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methoxy)(hydroxy)phosphoryl)oxy)oxidophosphoryl)oxy)-N,N,N-trimethylethanaminium

A178663-1G A178663-5G A178663-100MG A178663-1MG A178663-250MG A178663-25G
77600-444EA 19.48 USD
77600-444 77600-446 77625-838 77625-840 77625-842 77625-844
Citicoline ≥96%
Citicoline

Precaución: for Research Use Only

Formula: C₁₄H₂₆N₄O₁₁P₂
MW: 488.33 g/mol
MDL Number: MFCD00868097
CAS Number: 987-78-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR