Thietane-3-carboxylic acid 1,1-dioxide ≥95%, Ambeed.Inc

Supplier: Ambeed
Warning

A178601-100MG A178601-250MG A178601-1G
77611-732EA 293.55 USD
77611-732 77683-431 77890-140
Thietane-3-carboxylic acid 1,1-dioxide ≥95%, Ambeed.Inc
Thietane-3-carboxylic acid 1,1-dioxide

Precaución: For research use only.

Formula: C₄H₆O₄S
MDL Number: MFCD01859803
CAS Number: 13129-21-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR