tert-Butyl 4,7,10,13,16-pentaoxanonadec-18-yn-1-oate ≥95%

Supplier: Ambeed
Warning

Synonyms: Propargyl-PEG4-CH2CH2-Boc

A177824-1G A177824-250MG
77748-163EA 94 USD
77748-163 77748-164
tert-Butyl 4,7,10,13,16-pentaoxanonadec-18-yn-1-oate ≥95%
tert-Butyl 4,7,10,13,16-pentaoxanonadec-18-yn-1-oate

Precaución: for Research Use Only

Formula: C₁₈H₃₂O₇
MW: 360.44 g/mol
MDL Number: MFCD22201534
CAS Number: 1245823-50-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR