Erdafitinib ≥98%

Supplier: Ambeed
Warning

Synonyms: N1-(3,5-dimethoxyphenyl)-N2-isopropyl-N1-(3-(1-methyl-1H-pyrazol-4-yl)quinoxalin-6-yl)ethane-1,2-diamine, JNJ-42756493

A177503-1MG A177503-100MG A177503-250MG A177503-50MG
77615-490EA 11.33 USD
77615-490 77746-154 77746-155 77746-156
Erdafitinib ≥98%
Erdafitinib

Precaución: for Research Use Only

Formula: C₂₅H₃₀N₆O₂
MW: 446.55 g/mol
MDL Number: MFCD28502040
CAS Number: 1346242-81-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR