tert-Butyl 4-(hydroxymethyl)benzyl(methyl)carbamate ≥95%
Supplier: Ambeed
|
Warning
|
A177374-1G
A177374-100MG
A177374-250MG
A177374-5G
77593-132EA
188.6
USD
77593-132
77611-720
77611-722
77683-418
tert-Butyl 4-(hydroxymethyl)benzyl(methyl)carbamate ≥95%
tert-Butyl 4-(hydroxymethyl)benzyl(methyl)carbamate
Precaución: For research use only.
Formula:
C₁₄H₂₁NO₃ MW: 251.32 g/mol |
CAS Number:
774238-90-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...