SP2509 98+%

Supplier: Ambeed
Warning

Synonyms: (E)-N'-(1-(5-Chloro-2-hydroxyphenyl)ethylidene)-3-(morpholinosulphonyl)benzohydrazide, SP-2509

A177360-1MG A177360-5MG
77611-718EA 7.21 USD
77611-718 77685-398
SP2509 98+%
SP2509

Precaución: for Research Use Only

Formula: C₁₉H₂₀ClN₃O₅S
MW: 437.9 g/mol
MDL Number: MFCD28348370
CAS Number: 1423715-09-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR