5-Formyl-4-methylthiophene-2-boronic acid ≥98%

Supplier: Ambeed
Warning

A177274-1G A177274-250MG
77890-128EA 39 USD
77890-128 77890-129
5-Formyl-4-methylthiophene-2-boronic acid ≥98%
5-Formyl-4-methylthiophene-2-boronic acid

Precaución: for Research Use Only

Formula: C₆H₇BO₃S
MDL Number: MFCD03093890
CAS Number: 352530-25-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR