1-Oxa-9-azaspiro[5.5]undecane hydrochloride ≥95%

Supplier: Ambeed
Warning

A176783-100MG A176783-250MG A176783-1G
77598-232EA 73.8 USD
77598-232 77598-234 77748-157
1-Oxa-9-azaspiro[5.5]undecane hydrochloride ≥95%
1-Oxa-9-azaspiro[5.5]undecane hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₇NO·ClH
MDL Number: MFCD16556302
CAS Number: 1414958-73-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR