(2R,3R,4S,5S,6R)-2-(((3S,5R,6S,8R,9R,10R,12R,13R,14R,17S)-3,12-Dihydroxy-17-((S)-2-hydroxy-6-methylhept-5-en-2-yl)-4,4,8,10,14-pentamethylhexadecahydro-1H-cyclopenta[a]phenanthren-6-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol ≥98%

Supplier: Ambeed
Warning

Synonyms: Ginsenoside Rh1

A176502-100MG A176502-50MG
77748-155EA 97 USD
77748-155 77748-156
(2R,3R,4S,5S,6R)-2-(((3S,5R,6S,8R,9R,10R,12R,13R,14R,17S)-3,12-Dihydroxy-17-((S)-2-hydroxy-6-methylhept-5-en-2-yl)-4,4,8,10,14-pentamethylhexadecahydro-1H-cyclopenta[a]phenanthren-6-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol ≥98%
(2R,3R,4S,5S,6R)-2-(((3S,5R,6S,8R,9R,10R,12R,13R,14R,17S)-3,12-Dihydroxy-17-((S)-2-hydroxy-6-methylhept-5-en-2-yl)-4,4,8,10,14-pentamethylhexadecahydro-1H-cyclopenta[a]phenanthren-6-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol

Precaución: for Research Use Only

Formula: C₃₆H₆₂O₉
MDL Number: MFCD09951797
CAS Number: 63223-86-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR