4-Amino-2-hydroxybenzohydrazide ≥97%

Supplier: Ambeed
Warning

A176451-1G A176451-25G A176451-5G A176451-100MG A176451-250MG
77615-478EA 13.39 USD
77615-478 77615-480 77615-482 77748-153 77748-154
4-Amino-2-hydroxybenzohydrazide ≥97%
4-Amino-2-hydroxybenzohydrazide

Precaución: for Research Use Only

Formula: C₇H₉N₃O₂
MDL Number: MFCD01684522
CAS Number: 6946-29-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR