(R)-tert-Butyl 2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-3-iodopropanoate ≥98%

Supplier: Ambeed
Warning

Synonyms: Fmoc-3-Iodo-Ala-OtBu

A176240-100MG A176240-1G
77879-160EA 15 USD
77879-160 77879-161
(R)-tert-Butyl 2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-3-iodopropanoate ≥98%
(R)-tert-Butyl 2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-3-iodopropanoate

Precaución: for Research Use Only

Formula: C₂₂H₂₄INO₄
MDL Number: MFCD11053633
CAS Number: 282734-33-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR