3-Bromo-4,5-dimethylpyridine ≥98%

Supplier: Ambeed
Warning

A176108-1G A176108-250MG A176108-100MG A176108-5G
77671-500EA 101 USD
77671-500 77671-502 77871-882 77871-883
3-Bromo-4,5-dimethylpyridine ≥98%
3-Bromo-4,5-dimethylpyridine

Precaución: for Research Use Only

Formula: C₇H₈BrN
MDL Number: MFCD11656269
CAS Number: 27063-98-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR