6-Chloro-4-fluoro-1H-indole ≥97%

Supplier: Ambeed
Warning

A175734-100MG A175734-250MG
77890-125EA 30 USD
77890-125 77890-126
6-Chloro-4-fluoro-1H-indole ≥97%
6-Chloro-4-fluoro-1H-indole

Precaución: for Research Use Only

Formula: C₈H₅ClFN
MDL Number: MFCD07781560
CAS Number: 885520-88-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR