Azithromycin ≥98%

Supplier: Ambeed
Warning

Synonyms: (2R,3S,4R,5R,8R,10R,11R,12S,13S,14R)-11-(((2S,3R,4S,6R)-4-(Dimethylamino)-3-hydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)-2-ethyl-3,4,10-trihydroxy-13-(((2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyltetrahydro-2H-pyran-2-yl)oxy)-3,5,6,8,10,12,14-heptamethyl-1-oxa-6-azacyclopentadecan-15-one

A175409-100G A175409-10MG A175409-100MG A175409-25G A175409-500G A175409-50MG
77611-712EA 133.9 USD
77611-712 77611-714 77671-486 77671-488 77671-490 77671-492
Azithromycin ≥98%
Azithromycin

Precaución: for Research Use Only

MW: 749 g/mol
MDL Number: MFCD00873574
CAS Number: 83905-01-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR