3-[(tert-Butoxycarbonyl)amino]pyridine-2-carboxylic acid ≥95%

Supplier: Ambeed
Warning

Synonyms: 3-((tert-Butoxycarbonyl)amino)picolinic acid, 3-(Boc-amino)pyridine-2-carboxylic acid

A175372-1G A175372-250MG A175372-5G
77892-156EA 51 USD
77892-156 77892-157 77892-158
3-[(tert-Butoxycarbonyl)amino]pyridine-2-carboxylic acid ≥95%
3-[(tert-Butoxycarbonyl)amino]pyridine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₄N₂O₄
MW: 238.25 g/mol
MDL Number: MFCD02181061
CAS Number: 569687-82-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR