2,4,6-Trifluoro-3-hydroxyphenylboronic acid ≥98%, Ambeed.Inc

Supplier: Ambeed
Warning

A175338-100MG A175338-250MG A175338-1G
77683-408EA 40 USD
77683-408 77683-409 77885-366
2,4,6-Trifluoro-3-hydroxyphenylboronic acid ≥98%, Ambeed.Inc
2,4,6-Trifluoro-3-hydroxyphenylboronic acid

Precaución: For research use only.

Formula: C₆H₄BF₃O₃
MDL Number: MFCD11053885
CAS Number: 1072951-37-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR